Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
GraLab® Darkroom Timer, Dimco-Gray | Avantor

GraLab® Darkroom Timer, Dimco-Gray

120 V, 60 Hz
Catalog # 62373-007
Supplier:   Dimco-Gray
Fournisseur #:  7-300-160R
GraLab® Darkroom Timer, Dimco-Gray
Catalog # 62373-007
Supplier:  Dimco-Gray
GraLab® Darkroom Timer, Dimco-Gray
GraLab® Darkroom Timer, Dimco-Gray

120 V, 60 Hz

Price
$2,589.28
Log in pour la disponibilité
Each Case of 6
Product Details & Documents

Ideal for photographic, graphic arts, and printing darkroom use.

  • Provides 60 minutes of timing by minutes and seconds in one-second increments
  • Buzzer alarm with variable volume control signals end of time period

Black face has a single circle of interval time numerals. Hands and numerals are phosphorescent. With two three-wire outlet receptacles. Chemical- and moisture-resistant case is made from a blend of hybrid plastics.

Certifications: UL listed.

Spécifications
  • Electrical:
    120 V, 60 Hz
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.