Spécifications
- Cat. no.:62344-588
- W×H×D:6.4×8.6×2.2 cm (2¹/₂×3³/₈×⁷/₈")
Spécifications
A propos de cet article
Timer is the perfect reminder for signaling drying/incubating times, capturing test results, and alarming for appointments.
- Traceable®
- Triple display design
- Extra-loud, high-decibel alarm
Ideal for dedicating to tests that require several different times.
Three different countdown times may be set and then started simultaneously, or each channel may be started individually. Extra-loud, high-decibel alarm sounds for one minute or may be silenced manually.
Three-line display permits simultaneous countdown alarm timing of three different tests. Extra-large 1.3 cm (¹/₂") high display on each channel is easy to read from across the lab. Times may be set from 19 hours, 59 minutes, 59 seconds to 1 second. When zero is reached the timer starts counting up to show time elapsed since alarming. Additional features include stopwatch, time-out, and time-of-day. Magnetic back allows timer to be placed on any metal surface.