Order Entry
ContactUsLinkComponent
Bio-Safe II* Biodegradeable Cocktail
Bio-Safe II Complete Counting Cocktail, RPI
# de catalogue 61000-508
Fournisseur:  RPI
Small Business Enterprise
Numéro CAS:  
Bio-Safe II* Biodegradeable Cocktail
Bio-Safe II Complete Counting Cocktail, RPI
# de catalogue 61000-508
Fournisseur:  RPI
Small Business Enterprise
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    Bio-Safe II Complete Counting Cocktail
  • Size:
    4 L
  • Cat. no.:
    61000-508
  • Supplier No.:
    111195

Spécifications

A propos de cet article

High performance biodegradable cocktail for counting a wide range of samples including: aqueous solutions, biological materials, salt-buffer combinations and organic solutions.

Bio-Safe II has a sample holding capacity of up to 25% aqueous volume within a clear, stable region. This large capacity can actually reduce the amount of scintillation fluid required which becomes important when using miniature vials. Bio-Safe II will not permeate plastic scintillation vials even over extended storage periods.

Customers Who Bought This Also Bought