ContactUsLinkComponent
NALGENE* Polypropylene Peg Rack
Nalgene® Peg Racks, Polypropylene, Thermo Scientific
# de catalogue 60985-415
NALGENE* Polypropylene Peg Rack
Nalgene® Peg Racks, Polypropylene, Thermo Scientific
# de catalogue 60985-415
Spécifications
  • Length:
    73/8"
  • Width:
    4"
  • Color:
    White
  • Description:
    For O.D. Tubes
  • Height:
    23/4"
  • Cat. no.:
    60985-415
  • Supplier No.:
    5977-0017
Spécifications
A propos de cet article
Racks hold sample collection tubes, reaction tubes, and test tubes
Racks consist of an array of pegs spaced to hold test tubes upright and display the contents. Pegs are also excellent for holding inverted tubes for draining/drying.
Also hold and dry electrophoresis and chromatography plates. Durable, chemical-resistant, and autoclavable. Racks sink in water bath.
Customers Who Bought This Also Bought