Nalgene® Autoclaving Baskets, Polypropylene, Thermo Scientific
# de catalogue 60915-390
Fournisseur: Thermo Fisher Scientific
Spécifications
- Dimensions:15.2L x 12.4D x 10.5H cm (6 x 47/8 x 41/8")
- Cat. no.:60915-390
- Supplier No.:6917-0127
Spécifications
A propos de cet article
These baskets hold labware during autoclaving and washing
They allow complete venting and draining. Smooth surface will not scratch plastic or glass. White polypropylene will not rust. Nestable for storage.
Customers Who Bought This Also Bought