VWR® Bacti Cell Spreaders
Catalog # 60828-680
Supplier: VWR International
Specifications
- Sterility:Non sterile
- Color:Blue
- Cat. no.:60828-680
- Overall Length:21.5 cm
- Blade Width:30 mm
Specifications
About this item
Convenient and economical cell spreaders are designed to simplify bacterial culture work.
- Autoclavable
- Simplify bacterial culture work
- Durable fabrication
Durable enough to be autoclaved and reused.
Ordering information: Cell spreaders are available sterile (individually wrapped) or nonsterile (bulk packed).
Customers Who Bought This Also Bought