Black Phenolic Screw Caps for ACCUFORM® SSR™ Vials, Kimble Chase
# de catalogue 60826-315
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Cap Size:15-415
- Description:Phenolic Caps with PTFE-Faced/14B White Rubber Liner
- Cat. no.:60826-315
- Supplier No.:73802-15415