ContactUsLinkComponent
CAP 12MM PLUG NATURAL PK1000
Tiered Plug Stopper Closures, Polyethylene, Globe Scientific
# de catalogue 60820-140
Fournisseur:  Globe Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAP 12MM PLUG NATURAL PK1000
Tiered Plug Stopper Closures, Polyethylene, Globe Scientific
# de catalogue 60820-140
Fournisseur:  Globe Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Color:
    Natural
  • For Tubes:
    12 mm
  • Supplier No.:
    118127C
Spécifications
A propos de cet article
Polyethylene flanged plug caps provide exceptional leak-resistance in test tubes, centrifuge tubes, and round cuvettes
Closures fit 12mm tubes, and are available in a variety of colors to simplify identification and selection.
They feature two flexible flanges that press firmly against the inside of the tube wall, thereby providing better leak-resistance than standard plug caps.
Customers Who Bought This Also Bought