Hydrion® Double Roll pH Paper, Micro Essential Laboratory
Catalogue # 60778-041
Fournisseur: Micro Essential
Spécifications
- Description:pH Paper Dispenser, 3.0 - 8.0 pH Double Roll
- Cat. no.:60778-041
- Supplier No.:3-8
Spécifications
À propos de cet article
Hydrion® Double Roll pH Paper set the standard for quality, accuracy, and reliability.
Double Roll pH Papers consist of two rolls of pH papers packaged in a protective plastic dispenser with stainless steel cutoff lips and two matching color charts
Double Roll pH Wide Range Papers have a clear color match at each full pH unit: [0.0][1.0][2.0][3.0][4.0][5.0][6.0][7.0][8.0][9.0][10.0][11.0][12.0][13.0][14.0]
pH color changes are distinct and easily matched to the color chart.
Ordering information: Each carton (case) contains 10 dispensers, each consisting of two (2) 15-foot rolls of test paper and matching color charts. Provides for approximately 2000 tests.
Frequently Bought Together