ContactUsLinkComponent
Hydrion MicroFine 5.5-8.0 pH Paper Dispensers
Hydrion™, MicroFine pH Test Paper with Dispenser, Micro Essential Laboratory
# de catalogue 60775-964
Fournisseur:  Micro Essential
Hydrion™, MicroFine pH Test Paper with Dispenser, Micro Essential Laboratory
# de catalogue 60775-964
Fournisseur:  Micro Essential
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    pH Paper, Hydrion MicroFine Disp
  • Cat. no.:
    60775-964
  • pH Range:
    5.5 - 8.0
  • Supplier No.:
    MF-1606
Spécifications
A propos de cet article
This short-range Hydrion MicroFine pH Paper measures pH solutions in the acid pH range of 2.9-5.2 and offers clear bright single color matches at every 0.3 to 0.4 interval.
  • Provides precise, pinpoint pH measurement over a narrow pH range
  • Ultra-fast results with accuracy in intervals of 0.2 - 0.3 pH
  • Fit easily in to small tubes
These are ideally suited to testing minute amounts of solution, only requiring 0.0001mL for an accurate measurement.
Packaging: Packaged in a protective plastic dispenser with matching color chart contained in a polypropylene case to guard against sunlight and laboratory hazards.
Customers Who Bought This Also Bought