ContactUsLinkComponent
QT-10 Dispenser
Hydrion® Sanitizer Test Kits, Micro Essential
# de catalogue 60720-300
Fournisseur:  Micro Essential
Numéro CAS:  
QT-10 Dispenser
Hydrion® Sanitizer Test Kits, Micro Essential
# de catalogue 60720-300
Fournisseur:  Micro Essential
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Packaging:
    Contains dispenser, color chart and 1 roll of test paper, enough for 100 tests or more.
  • Description:
    QT-10 Quaternary Ammonium Test Kit, 0-400 ppm
  • Range:
    0-400 ppm
  • Supplier No.:
    QT-10
Spécifications
A propos de cet article
Use Hydrion Sanitizer test papers wherever food is commercially produced or served to test the concentration of sanitizers, as required by health code
Ammonium test kit and kit refill are for testing first and second generation Quats. Match points at 0, 100, 200, 300, and 400ppm.
Chlorine test kit and kit refill are for testing free chlorine in sanitizing rinse. Match points at 0, 50 100, and 200ppm.
These test papers provide a simple, reliable way to meet Federal, State, and local food safety guidelines
Customers Who Bought This Also Bought