SP Bel-Art FLUO-KEM® Teflon® Resin Lab-Thread Tape, Bel-Art Products, a part of SP
Catalog # 60490-100
Supplier: Bel-Art Products, a Part of SP
Specifications
- Width:1.3 cm (½")
- Length:7.6 m (300")
- Cat. no.:60490-100
- Supplier No.:240200000
Specifications
About this item
Pure, non-sintered Teflon® PTFE Lab-Thread Tape contains no adhesive, yet does a superior job of sealing small apertures.
- Non-sintered
- Seals small apertures securely without adhesive
- Withstands a wide range of temperatures
- No need for lubricants and vacuum greases
Conforming to irregularities of surfaces, threads and joints of all types, it eliminates the need for lubricants and vacuum greases
Can be used on ground glass joints, desiccators, bell jars, freezedrying equipment, etc. 0.0635 mm (0.0025") thick, 12.7 mm (½") wide x 7.6 meters long (300") per roll. Withstands temperatures from -250ºF to 500ºF.
Provides a vacuum tight seal and eliminates ˝freezing˝ even after long periods.
Customers Who Bought This Also Bought