Order Entry
ContactUsLinkComponent
VWR* Talon* Rods, Fiberglass
VWR® Talon® Rods, Fiberglass
# de catalogue 60079-523
Fournisseur:  VWR International
VWR* Talon* Rods, Fiberglass
VWR® Talon® Rods, Fiberglass
# de catalogue 60079-523
Fournisseur:  VWR International

Spécifications

  • Rod Length:
    610 mm (24")
  • Cat. no.:
    60079-523

Spécifications

A propos de cet article

Fiberglass rods are structurally strong and resistant to clamp compression scars.

  • Strong and resilient
  • For use with VWR® Talon® lab frames
  • Resistant to corrosion and rust
  • Fiberglass construction

Diameter: 13mm (1/2").

Customers Who Bought This Also Bought