VWR® Rubber Stoppers
# de catalogue 59586-163
Fournisseur: VWR International
Small Business Enterprise
Spécifications
- Size:9
- Cat. no.:59586-163
Spécifications
A propos de cet article
Versatile rubber stoppers are four stoppers in one.
- Easy conversion between stopper types
- Versatile design
- Soft and resilient
- Natural rubber construction
- Autoclavable
Three simple twist-off seals permit easy conversion from a solid to a one, two, or three-hole stopper for all your laboratory needs. Soft, resilient, natural, white rubber demonstrates exceptional aging characteristics and withstands repeated autoclaving.
Ordering information: Stoppers are available in sizes 3 to 10 or as an assortment of 33 stoppers. Sizes 3 and 4 only have two twist-off seals. Stopper assortment 59586-167 contains nine size 3 stoppers, eight size 4, six size 5, four size 6, two size 7, two size 8, one size 9, and one size 10 stopper. For black stoppers (shown above), contact your Avantor representative.
Customers Who Bought This Also Bought