PYREX® Polyethylene [ST] Stoppers, Corning
Catalog # 59492-123
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Cat. no.:59492-123
- ST Stopper No.:27
- Supplier No.:7624-27
Specifications
About this item
These PYREX® No.13 stoppers are made from linear HDPE and have closed bottoms to conform to Standard Taper stopper dimensions.
- Blue and white with a closed bottom
- Three rings on the plug provide an efficient seal
- Improved design fully protects the neck from accidental tipping
Frequently Bought Together