ContactUsLinkComponent
PYREX brand HDPE Polyethylene, Improved Form, Stoppers, Caps and Closures
PYREX® Polyethylene [ST] Stoppers, Corning
Catalog # 59492-123
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
undefined
PYREX® Polyethylene [ST] Stoppers, Corning
Catalog # 59492-123
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
Specifications
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Cat. no.:
    59492-123
  • ST Stopper No.:
    27
  • Supplier No.:
    7624-27
Specifications
About this item
These PYREX® No.13 stoppers are made from linear HDPE and have closed bottoms to conform to Standard Taper stopper dimensions.
  • Blue and white with a closed bottom
  • Three rings on the plug provide an efficient seal
  • Improved design fully protects the neck from accidental tipping
Frequently Bought Together