Nalgene® Stirring Rod, Polypropylene, Thermo Scientific
Catalog # 59062-097
Supplier: Thermo Fisher Scientific
Specifications
- Description:Polypropylene Stirring Rod
- Cat. no.:59062-097
- Supplier No.:6169-0010
Specifications
About this item
Versatile stirring rod with a flat, flexible, 15.9x15.9mm (5/8x5/8") paddle at one end, and a triangular paddle at other end
Length: 247.7mm (93/4"). Combines functions of stirring rod and rubber policeman.
Customers Who Bought This Also Bought