Order Entry
ContactUsLinkComponent
VWR* Spin-Wedge* Stir Bars
VWR® Spinwedge® Magnetic Stir Bars
# de catalogue 58949-265
Fournisseur:  VWR International
VWR* Spin-Wedge* Stir Bars
VWR® Spinwedge® Magnetic Stir Bars
# de catalogue 58949-265
Fournisseur:  VWR International

Spécifications

  • Coating:
    PTFE
  • Ø:
    50 mm (2")
  • Length:
    12 mm (½")
  • Cat. no.:
    58949-265
  • Shape:
    Wedge

Spécifications

A propos de cet article

Spinwedge® magnetic stirring bars provide strong turbulence at fairly low speeds and are well suited for churning sediment or dissolving salts

  • Wedge shape is designed to plow up sediment for thorough dispersion
  • Sharply angled broad face provides strong mixing action, perfect for viscous solutions
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Customers Who Bought This Also Bought