Order Entry
Canada
ContactUsLinkComponent
VWR® Spinbar® Magnetic Stir Bars, Egg-Shaped
Catalogue # 58949-243
Fournisseur:  VWR International
undefined
VWR® Spinbar® Magnetic Stir Bars, Egg-Shaped
Catalogue # 58949-243
Fournisseur:  VWR International

Spécifications

  • Coating:
    PTFE
  • Ø:
    19 mm (3/4 ")
  • Length:
    64 mm (2½")
  • Cat. no.:
    58949-243
  • Shape:
    Egg-shaped
  • For Use With:
    3000, 5000 ml Flasks

Spécifications

À propos de cet article

These egg-shaped stirring bars, designed especially for use in round-bottom vessels, spin unrestricted by the vessel contour.

  • Excellent for round-bottom flasks
  • Provide their own pivot point without a separate ring or abrupt change in contour
  • Offers minimal contact when used in plastic containers

The magnet itself is egg-shaped with a Teflon® PTFE coating, except the three smallest sizes have a cylindrical magnet in an egg-shaped Teflon® PTFE coating. Autoclavable.

Frequently Bought Together