VWR® Spinbar® Micro Stir Bars
# de catalogue 58948-353
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Coating:PTFE
- Ø:1.5 mm (1/16 ")
- Length:7.9 mm (5/16")
- Description:Stir Bar
- Cat. no.:58948-353
- Shape:Polygon
Spécifications
A propos de cet article
Micro (Flea) magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes, and gradient makers.
- Particularly useful for environmental testing
- Valuable for preparing and evaluating small sample volumes
- Teflon® PTFE encapsulated
- Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Customers Who Bought This Also Bought