Order Entry
ContactUsLinkComponent
VWR* Spinplus* Stir Bars
VWR® Spinplus® Magnetic Stir Bars
# de catalogue 58947-826
Fournisseur:  VWR International
VWR* Spinplus* Stir Bars
VWR® Spinplus® Magnetic Stir Bars
# de catalogue 58947-826
Fournisseur:  VWR International

Spécifications

  • Coating:
    PTFE
  • Ø:
    32 mm (11/4")
  • Length:
    32 mm (11/4")
  • Description:
    Spin Bar
  • Height:
    14.3 mm (9/16")
  • Cat. no.:
    58947-826
  • Shape:
    Cross-shaped

Spécifications

A propos de cet article

Spinplus® magnetic stirring bars add speed and efficiency to mixing operations.

  • The “+” shape creates a deep vortex and provides stable, quiet operation
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable

Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low-friction coating is durable and inert.

Ordering information: Order each size separately or choose a five-piece assortment (one of each size).
Customers Who Bought This Also Bought