VWR® Fine-Pore Cellulose Sponges
Catalog # 58540-069
Supplier: VWR International
Specifications
- Dimensions:Expanded: 15.2 x 11.4 x 2.5 cm (6 x 41/2 x 1")
- Cat. no.:58540-069
Specifications
About this item
These fine-pore natural color cellulose sponges hold about 25 times their own weight in water.
- Durable
- Convenient for quick laboratory wipe-up
- Resist tearing and abrasion
- Absorb liquid more evenly than large pore sponges
Available in compressed (approximately ¹/₈" thickness before expansion) or uncompressed format.
Frequently Bought Together