ContactUsLinkComponent
VWR* Cellulose Sponges, Fine Pore
VWR® Fine-Pore Cellulose Sponges
Catalog # 58540-069
Supplier:  VWR International
undefined
VWR® Fine-Pore Cellulose Sponges
Catalog # 58540-069
Supplier:  VWR International
Specifications
  • Dimensions:
    Expanded: 15.2 x 11.4 x 2.5 cm (6 x 41/2 x 1")
  • Cat. no.:
    58540-069
Specifications
About this item
These fine-pore natural color cellulose sponges hold about 25 times their own weight in water.
  • Durable
  • Convenient for quick laboratory wipe-up
  • Resist tearing and abrasion
  • Absorb liquid more evenly than large pore sponges
Available in compressed (approximately ¹/₈" thickness before expansion) or uncompressed format.
Frequently Bought Together