VWR® Fine-Pore Cellulose Sponges
Catalog # 58540-047
Supplier: VWR International
Specifications
- Dimensions:Expanded: 14 x 9.2 x 2.5 cm (51/2 x 35/8 x 1")
- Cat. no.:58540-047
Specifications
About this item
These fine-pore natural color cellulose sponges hold about 25 times their own weight in water.
- Durable
- Convenient for quick laboratory wipe-up
- Resist tearing and abrasion
- Absorb liquid more evenly than large pore sponges
Available in compressed (approximately ¹/₈" thickness before expansion) or uncompressed format.
Customers Who Bought This Also Bought