SP Bel-Art Vikem® Pipette Filler,Vinyl, Bel-Art Products, a part of SP
Catalog # 57497-306
Supplier: Bel-Art Products, a Part of SP
Specifications
- Ø:5 cm
- Description:Pipette Filler
- Length:10 cm
- Cat. no.:57497-306
- Supplier No.:378860000
Specifications
About this item
This pipette aid is operated by squeezing then releasing the bulb to aspirate fluid into the pipet.
- Vikem® vinyl with a tapered polyethylene chuck
- Designed for quick removal when filling is complete
- Simple to use
After aspirating the fluid, the pipette aid is then removed, and the pipette top quickly covered with a gloved finger to control and dispense fluid.
Customers Who Bought This Also Bought