Spécifications
- Dimensions:30.5Lx30.5Wx68.5H cm (12x12x27)"
- Description:Floor disposal carton
- Cat. no.:56617-801
Spécifications
A propos de cet article
Disposal cartons provide a safe, convenient place to throw away broken glassware and residual liquids.
Disposable cardboard receptacles with 2mil thick polypropylene liner safely contain broken glass and residual liquids. Entire unit may be disposed of as a whole once safety cap is pulled into place. Available in floor and benchtop models.
Optional, reusable carton cover (for floor model only) provides a wider opening 20.3 x 24.1cm (8 x 9.5”) permitting safer and more sanitary deposit of wastes into the container. Opens quickly and easily by pressing the built-in release tab - no batteries required! Features a smooth 10.1 x 10.1cm (4 x 4”) area on top for applying labels. Autoclavable lid is color-coded for easy identification.
- Easy to assemble
- Disposable cardboard construction
- Safe and convenient disposal of broken glassware
- Color-coded lids
The entire unit can be disposed of as a whole once the safety cap is pulled into place. The autoclavable lid is color-coded for easy identification, and the cartons are available in floor and benchtop models. Optional reusable carton covers, for the floor model only, provide a wider opening of 20.3×24.1 cm (8×9.5") for safer and more sanitary deposit of wastes into the container. The included polypropylene bag is 2 mil thick.
Pressing the built-in release tab opens the carton. No batteries are required. The carton features a smooth area on top for applying labels.