ContactUsLinkComponent
VWR* Absorbent Bench Underpad; Underpads; Under Pads; Pads; Absorbent Pads; Bench Pads; Polypropylene Pads; Cellulose Pads; Polymer Pads; Sealed Pads; Wicking Pads; Cushion Pads; Deluxe Pads; Spunbound Pads; Spill Pads; Bench Covers; Chux; Chucks; Bedmats; Bed Mats; Incontinence; Incontinent
VWR® Absorbent Underpads with Waterproof Moisture Barrier
# de catalogue 56617-000
Fournisseur:  VWR International
VWR* Absorbent Bench Underpad; Underpads; Under Pads; Pads; Absorbent Pads; Bench Pads; Polypropylene Pads; Cellulose Pads; Polymer Pads; Sealed Pads; Wicking Pads; Cushion Pads; Deluxe Pads; Spunbound Pads; Spill Pads; Bench Covers; Chux; Chucks; Bedmats; Bed Mats; Incontinence; Incontinent
VWR® Absorbent Underpads with Waterproof Moisture Barrier
# de catalogue 56617-000
Fournisseur:  VWR International
Spécifications
  • W×L:
    58.4 x 91.4 cm (23 x 36")
  • Description:
    Deluxe underpads, blue backing
  • Absorbency:
    600 ml
Spécifications
A propos de cet article
These super absorbent underpads feature a multi-layer, cushioned construction that is breathable yet absorbent enough for hospital and laboratory use. The polymer layer channels and blocks fluids in the core for maximum fluid retention while eliminating odors and wicking wetness away from the skin.
  • Three-layers of absorbent material
  • Soft white spunbound polypropylene top sheet
  • Super absorbent polymer made of cellulose
  • Sealed on four sides to prevent liquid seepage
These pads are ideal for covering hospital beds and chairs, as well as for use during IV starts and minor surgical procedures
When used as a laboratory bench pad or bench cover, these underpads serve as a protective barrier for labware to prevent breakage and collect spills. Extra large sizes are available, to provide maximum coverage and protection. The backsheets are color-coded (blue or green). Deluxe pads are more absorbent and thicker with more absorbent celluose fluff than standard pads. Super deluxe pads have an additional super absorbent polymer.
Customers Who Bought This Also Bought