Nalgene® Safety Bottle Carriers, Thermo Scientific
# de catalogue 56609-222
Fournisseur: Thermo Fisher Scientific
Spécifications
- Capacity:4 L (1 gal.)
- Description:LDPE safety bottle carrier
- Cat. no.:56609-222
- Supplier No.:6501-4000
Spécifications
A propos de cet article
For the transport and storage of stockroom chemicals, glass bottles, and field samples
Clear plastic lid permits identification of color-coded closures on acid bottles and protects the neck and bottle cap from damage. Lid may be removed with a quick twist; pour directly from the bottle without removal from the carrier.
Carriers are manufactured of impact-resistant, low-density polyethylene and hold metric-sized chemical bottles for improved safety and ease of handling. Bottles with fingerloops fit easily.
Customers Who Bought This Also Bought