ContactUsLinkComponent
Nalgene Safety Bottle Carriers, Thermo Scientific
Nalgene® Safety Bottle Carriers, Thermo Scientific
# de catalogue 56609-222
Nalgene Safety Bottle Carriers, Thermo Scientific
Nalgene® Safety Bottle Carriers, Thermo Scientific
# de catalogue 56609-222
Spécifications
  • Capacity:
    4 L (1 gal.)
  • Description:
    LDPE safety bottle carrier
  • Cat. no.:
    56609-222
  • Supplier No.:
    6501-4000
Spécifications
A propos de cet article
For the transport and storage of stockroom chemicals, glass bottles, and field samples
Clear plastic lid permits identification of color-coded closures on acid bottles and protects the neck and bottle cap from damage. Lid may be removed with a quick twist; pour directly from the bottle without removal from the carrier.
Carriers are manufactured of impact-resistant, low-density polyethylene and hold metric-sized chemical bottles for improved safety and ease of handling. Bottles with fingerloops fit easily.
Customers Who Bought This Also Bought