Nalgene® Safety Bottle Carriers, Thermo Scientific
# de catalogue 56609-200
Fournisseur: Thermo Fisher Scientific
Spécifications
- Capacity:2.5 L (5 pt.)
- Description:LDPE safety bottle carrier, LDPE
- Cat. no.:56609-200
- Supplier No.:6501-2500
Spécifications
A propos de cet article
For the transport and storage of stockroom chemicals, glass bottles, and field samples
Clear plastic lid permits identification of color-coded closures on acid bottles and protects the neck and bottle cap from damage. Lid may be removed with a quick twist; pour directly from the bottle without removal from the carrier.
Carriers are manufactured of impact-resistant, low-density polyethylene and hold metric-sized chemical bottles for improved safety and ease of handling. Bottles with fingerloops fit easily.
Customers Who Bought This Also Bought