Order Entry
ContactUsLinkComponent
VWR* Bottle Tote* Small Safety Carrier
VWR® Bottle Tote™ Safety Carriers
# de catalogue 56608-982
Fournisseur:  VWR International
VWR* Bottle Tote* Small Safety Carrier
VWR® Bottle Tote™ Safety Carriers
# de catalogue 56608-982
Fournisseur:  VWR International

Spécifications

  • Ø:
    11.4 cm (4½")
  • Color:
    Black
  • Description:
    Bottle tote, small, 500 ml bottle + 1 L bottle
  • Cat. no.:
    56608-982

Spécifications

A propos de cet article

Carriers are designed for the safe transport of acids, alkalies, and solvents.

  • High degree of tensile strength
  • Centering ring assures proper clearance for safe transport
  • Molded-in handle

High in tensile strength, carriers can be used in temperatures from -45 to 110°C (-49 to 230°F) and can be steam sterilized

The small carriers can accommodate 500 mL (16.9 oz.) or 1L (33.8 oz.) bottles. The large carriers can accommodate 2.5L (84.5 oz.) or 4L (135 oz.) bottles.

Centering ring in base assures a 2.54 cm (1") clearance between bottle and wall of carrier. Molded-in handle gives a comfortable grip.

Customers Who Bought This Also Bought