VWR® Neoprene Flask Underlay
# de catalogue 56255-006
Fournisseur: VWR International
Spécifications
- Ø:13 cm (51/8")
- Cat. no.:56255-006
Spécifications
A propos de cet article
Moderate heat and oil resistance with concave recess on top to support round-bottom flasks and dishes.
- Made of long-lasting green Neoprene rubber
Holds 250 and 500 mL flasks firmly in place.
Customers Who Bought This Also Bought