PUMP UNMOUNTED 1380
# de catalogue 54987-200
Fournisseur: Welch Vacuum
Customers Who Bought This Also Bought
Sodium hydroxide 50% (w/w) in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
220,41 $ to 875,50 $
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Fournisseur: VWR International
881,80 $ to 2 807,56 $
EDTA disodium salt concentrate for dilution to 1L, 0.1 M EDTA-Na2 volumetric solution, for complexometry, Fluka™
Fournisseur: Honeywell Research Chemicals
153,96 $
Sodium azide ≥99.7%, OR, Macron Fine Chemicals™
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
350,13 $
Hydrochloric acid 5 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
292,27 $ to 912,91 $