Order Entry
Canada
ContactUsLinkComponent
SP Bel-Art Aquet® Detergent for Glassware and Plastics, Bel-Art Products, a part of SP
Catalogue # 54848-004
undefined
SP Bel-Art Aquet® Detergent for Glassware and Plastics, Bel-Art Products, a part of SP
Catalogue # 54848-004
Supplier Number:  F17094-0050

Spécifications

  • Description:
    Cubitainer® container
  • Size:
    19 L (5 gal.)
  • Cat. no.:
    54848-004
  • Supplier No.:
    170940050

Spécifications

À propos de cet article

Safe for glassware, plastics, and surfaces, this phosphate-free, nonionic surfactant will not etch or mar precision glassware.

  • Convenient, pre-portioned pouches make one gallon of 1% solution
  • 3.8 L (1 gal.) bottle comes with 473 ml (16 oz.) working solution dispenser bottle
  • 20 L (5 gal.) Cubitainer™ is available for frequent users
  • Not for use in automatic dishwashers

It rinses quickly and dries without wiping.

Frequently Bought Together