Order Entry
ContactUsLinkComponent
NALGENE* Pipet Washer-Rinser
Nalgene® Pipette Washer-Rinsers, Thermo Scientific
# de catalogue 53595-020
NALGENE* Pipet Washer-Rinser
Nalgene® Pipette Washer-Rinsers, Thermo Scientific
# de catalogue 53595-020

Spécifications

  • For:
    Pipettes up to 40.6 cm (16") length
  • Cat. no.:
    53595-020
  • Overall Height:
    77.5 cm (30½")
  • Supplier No.:
    5245-0040

Spécifications

A propos de cet article

One-piece, high-density polyethylene washer-rinser is leakproof.

Water inlet at the top prevents back-siphoning of contaminated water. Operates at wide range of cycling speeds, with water input from 1.5 to 12 L per minute.

Ordering information: Includes inlet and outlet tubing ready for immediate installation. Compatible pipette basket is available separately (see Accessories tab below).
Customers Who Bought This Also Bought