MµlTI™ SafeSeal™ Microcentrifuge Tubes, Sorenson BioScience
# de catalogue 53550-698
Fournisseur: Sorenson Bioscience
Spécifications
- Sterility:Non sterile
- Volume:1.7 ml
- Color:Natural
- Description:Tube
- Cat. no.:53550-698
- Maximum RCF:30000×g
- Supplier No.:16070
Spécifications
A propos de cet article
These disposable microtubes are designed for use in all popular microcentrifuges.
These disposable microtubes are designed for use in all popular microcentrifuges.
They are manufactured from precision molded polypropylene for biotechnology applications. The cap design allows repeated openings and closings yet resists popping from internal pressure or during boiling.
Delivery: Tubes 53550-915 are supplied in individual bags of 200 of blue, yellow, violet, green, and red. Tubes 53550-778 are supplied in individual bags of 100 of blue, yellow, violet, green, and red. Tubes 53550-870 are supplied in individual bags of 80 of blue, yellow, violet, green, and red.
Customers Who Bought This Also Bought