ContactUsLinkComponent
Finntip* Pipet Tips
Finntip™ Pipette Tips, Thermo Scientific
# de catalogue 53516-150
Finntip™ Pipette Tips, Thermo Scientific
# de catalogue 53516-150
Spécifications
  • Volume:
    0.5 - 250 µl
  • Packed:
    1000/Bag
  • Tip Style:
    Finntip™ 250 Universal
  • Supplier No.:
    9400230
Spécifications
A propos de cet article
These high-quality pipette tips are specifically designed to fit Finnpipette® pipettors, but are also compatible with most pipettor brands.
  • Made of high-quality polypropylene
  • Autoclavable
  • Chemical-resistant
Tips in refill trays can be used to quickly refill existing racks.
Customers Who Bought This Also Bought