ContactUsLinkComponent
VWR* PCR Tube Strips with Cap Strips
VWR® PCR 12-Well Tube Strips and Caps
# de catalogue 53509-306
Fournisseur:  VWR International
   Sustainable Packaging
   Low Manufacturing Impact
Product available on GSA Advantage®
VWR* PCR Tube Strips with Cap Strips
VWR® PCR 12-Well Tube Strips and Caps
# de catalogue 53509-306
Fournisseur:  VWR International
   Sustainable Packaging
   Low Manufacturing Impact
Product available on GSA Advantage®
Spécifications
  • Cap Type:
    Domed
  • Capacity:
    0.2 ml
  • Format:
    Strip
  • Color:
    Natural
  • Description:
    Tube Strip with Separate Bubble Cap Strip
  • Environmentally Preferable:
    Sustainable Packaging,
    Low Manufacturing Impact
Spécifications
A propos de cet article
Ideal for all PCR protocols, these 0.2 ml tubes and caps are made from polypropylene and are compatible with most popular thermal cycler blocks.
  • Chemically inert
  • Autoclavable
  • Caps ensure a contamination-free environment
They feature smooth, thin, uniform walls for accurate thermal transfer. Certified free of RNase, DNase, and human DNA (lot-by-lot certificates available). Caps fit precisely.
Ordering information: Tube strips with separate cap strips (53509-306) are packed with 80 tubes and 80 caps in separate bags inside a box.
Customers Who Bought This Also Bought