VWR® PCR 12-Well Tube Strips and Caps
# de catalogue 53509-306
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Cap Type:Domed
- Capacity:0.2 ml
- Format:Strip
- Color:Natural
- Description:Tube Strip with Separate Bubble Cap Strip
- Environmentally Preferable:Sustainable Packaging,Low Manufacturing Impact
Spécifications
A propos de cet article
Ideal for all PCR protocols, these 0.2 ml tubes and caps are made from polypropylene and are compatible with most popular thermal cycler blocks.
- Chemically inert
- Autoclavable
- Caps ensure a contamination-free environment
They feature smooth, thin, uniform walls for accurate thermal transfer. Certified free of RNase, DNase, and human DNA (lot-by-lot certificates available). Caps fit precisely.
Ordering information: Tube strips with separate cap strips (53509-306) are packed with 80 tubes and 80 caps in separate bags inside a box.
Customers Who Bought This Also Bought