Pure™ 1000 SoftFit™ Pipette Tips, Molecular BioProducts
Catalog # 53225-708
Supplier: Thermo Scientific
Specifications
- Volume:100 - 1000 µL
- Length:7.9 cm (3⅛")
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
- Supplier No.:3580
Specifications
About this item
The SoftFit design of these tips ensures tip-to-tip precision and universal fit.
- Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
- Nonsterile tips are autoclavable
This feature also makes loading and ejecting easy and minimizes hand fatigue. Tips are compatible with most major brand pipettors.
Ordering information: Tips 53225-712 and -710 are packaged 100 per tray, 8 trays per pack. Tips 53225-708 are bulk packaged.
Frequently Bought Together