Order Entry
ContactUsLinkComponent
Pure* 10 Pipet Tips
Pure™ 10 Pipette Tips, Molecular BioProducts
# de catalogue 53225-662
Fournisseur:  Thermo Scientific
Pure™ 10 Pipette Tips, Molecular BioProducts
# de catalogue 53225-662
Fournisseur:  Thermo Scientific

Spécifications

  • Length:
    3.14 cm (1.236")
  • Volume:
    0.1 - 10 µl
  • Supplier No.:
    3502

Spécifications

A propos de cet article

These pipette tips are ideal for low volume pipetting.

  • Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
  • Nonsterile tips are autoclavable

The MicroPoint* design increases precision when working with low volume samples as the tip profile is nearly 50% smaller than standard blunt-end tips and much more narrow than beveled-end tips. Tips are ideal for a wide variety of applications and are compatible with Pipetman*, Nichiryo*, Biohit*, Finnpipette*, Rainin*, and Transferpette* pipettors.

Ordering information: Tips 53225-664 and -662 are packaged 96 per tray, 10 trays per pack. Tips 53225-660 are bulk packaged.
Customers Who Bought This Also Bought