Flipper Racks, 4-Way, Thermo Scientific
# de catalogue 53225-626
Fournisseur: Thermo Scientific
Spécifications
- Color:Fluorescent Green
- Cat. no.:53225-626
- Supplier No.:8850
Spécifications
A propos de cet article
Thermo Scientific Molecular BioProducts 4-Way Flipper Rack holds thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side.
Customers Who Bought This Also Bought