ContactUsLinkComponent
FLIPPER rack, 96-well, with lid
96-Well Flipper™ Microtube Racks, Molecular BioProducts
# de catalogue 53225-610
Fournisseur:  Thermo Scientific
FLIPPER rack, 96-well, with lid
96-Well Flipper™ Microtube Racks, Molecular BioProducts
# de catalogue 53225-610
Fournisseur:  Thermo Scientific
Spécifications
  • Color:
    Fluorescent Pink
  • Description:
    96-Well Flipper Rack
  • Material:
    PP
  • Supplier No.:
    8760
Spécifications
A propos de cet article
Protect samples and allow racks to be stacked using this 96-well flipper microtube rack.
  • Holds 0.5 mL tubes on one side, 1.5 mL tubes on the other
  • Molded with an alphanumeric grid for easy tube identification
  • Sides have a frosted writing surface for smear-resistant labeling
  • Several colors available
Customers Who Bought This Also Bought