Nalgene® Marking Pens, Thermo Scientific
# de catalogue 52887-030
Fournisseur: Thermo Fisher Scientific
Spécifications
- Point Type:Bold
- Color:Black
- Description:Marking pens
- Cat. no.:52887-030
- Supplier No.:6311-0010
Spécifications
A propos de cet article
Permanent black ink, pocket clip felt-tip pens for writing on glass and plastic surfaces
- Markings will not be affected by autoclaving
Customers Who Bought This Also Bought