ContactUsLinkComponent
Sight Savers® Premoistened Lens Cleaning Tissues, Bausch & Lomb®
Catalogue # 52848-078
Fournisseur:  Bausch & Lomb
undefined
Sight Savers® Premoistened Lens Cleaning Tissues, Bausch & Lomb®
Catalogue # 52848-078
Fournisseur:  Bausch & Lomb
Supplier Number:  8574GM
Spécifications
  • Description:
    Premoistened Lens Cleaning Tissues
  • Cat. no.:
    52848-078
  • Supplier No.:
    8574GM
Spécifications
À propos de cet article
These nonsilicone, antistatic tissues are individually packaged and treated with special surfactants to clean all glass and plastic lenses.
Perfect for computer screens, mirrors, safety glasses, and face shields. Unique package design allows easy dispensing of large 12.7×20.3 cm (5×8") folded, premoistened tissues. Cardboard dispenser box measures 22.9×13.7×6 cm (9×5³/₈×2³/₈").
Frequently Bought Together