Nalgene® Polyolefin Plastic Paper Sheets, Thermo Scientific
# de catalogue 51280-114
Fournisseur: Thermo Fisher Scientific
Spécifications
- Cat. no.:51280-114
- Dimensions:8.5×11"
- Supplier No.:6304-9811
Spécifications
A propos de cet article
Nalgene® Polyolefin Plastic Paper Sheets roll or fold without cracking and stay flexible at cold temperatures
They won't rip, stretch or shrink and can be punched for use in binders or cut into labels. This paper is resistant to many solvents including acetone, formaldehyde, formalin, xylene, acetic acid and ethanol. They accept many inks including laser, inkjet, copier, markers, ballpoint, gels and technical markers. These sheets DO NOT accept pencil.
Customers Who Bought This Also Bought