SP Bel-Art Slide Mailer, Bel-Art Products, a part of SP
# de catalogue 48428-029
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Description:Slide Mailer
- Cat. no.:48428-029
- Supplier No.:442000000
Spécifications
A propos de cet article
Chemically inert polyethylene container includes a deep screw cap to provide maximum protection against breakage.
- Polyethylene construction
- Chemically inert
- Easy to grip and clean when projected out of jar
Once the cap is removed, slides project 12.7 mm (½") out of the jar for easy grip and clean handling. Internal grooves accept 5 to 10 microscope 25x75 mm (1x3") slides, or can be used as a specimen jar to hold vials up to 12 mL. The cap has a small loop for attachment of wired tags and should be taped for shipment.
Customers Who Bought This Also Bought