SP Bel-Art Coplin Staining Jar, Bel-Art Products, a part of SP
Catalog # 47751-792
Supplier: Bel-Art Products, a Part of SP
Specifications
- Dimensions:5.9 Dia. x 11.4 H cm (25/16 x 41/2")
- Cat. no.:47751-792
- Supplier No.:442081000
Specifications
About this item
This polypropylene staining jar improves on the classic tapered square design with a larger opening for easier placement and removal of slides.
- Polypropylene construction
- Larger opening than classic tapered square jars
- Easy to place and remove slides
- Autoclavable
The polypropylene screw cap is 53 mm in diameter. It has a flat top, easy-grip large knurled edge, deep thread, and a seal ring to prevent leaks. Five internal slots hold up to 10 standard 25x75 mm or 1x3" slides back to back. Jar is steam autoclavable at 121°C (250°F). Remove cap when autoclaving.
Customers Who Bought This Also Bought