ContactUsLinkComponent
MAXYMum Recovery* Universal Fit Pipet Filtered Tip
Axygen® MAXYMum Recovery™ Hydrophobic Filter Barrier Pipette Tips, Corning
Catalog # 47749-868
Supplier:  Corning
Product available on GSA Advantage®
Axygen® MAXYMum Recovery™ Hydrophobic Filter Barrier Pipette Tips, Corning
Catalog # 47749-868
Supplier:  Corning
Product available on GSA Advantage®
Specifications
  • Volume:
    0.5 - 10 µL
  • Cat. no.:
    47749-868
  • Tip Style:
    Extended length
  • For Use With:
    Most Research-Grade Pipettors
  • Supplier No.:
    TXLF-10-L-R-S
Specifications
About this item
For applications that require high accuracy and reproducibility.
  • Compatible with most major brands of pipettors
Manufactured using diamond-polished molds to eliminate the surface cavities and occlusions which cause sample retention. Polymer resists DNA, RNA, and protein adherence and will not denature DNA. Manufactured without secondary substances such as silicone or heavy metals, which can contaminate samples.
Tips are autoclavable and presterilized through exposure to e-beam radiation for protection from contamination.
Ideal for DNA multiplication and handling radioactive, biohazardous, and corrosive samples. Filter prevents cross-contamination from the pipette to subsequent samples.
Certifications: Certified to be free of DNase, RNase, and pyrogens.
Ordering information: The 10 to 300 µl tips and 1250 µl tips 12000-756 are packaged 96 per sealed-bottom hinged rack, 500 and 1000 µl tips are packaged 100 per non-hinged rack. For non sterile tips, contact your sales representative.
Customers Who Bought This Also Bought