Axygen® Sequencing Septa Mats, Corning
Catalog # 47749-866
Supplier: Corning
Specifications
- Description:Sequencing Septa Mat
- Material:Silicone
- Cat. no.:47749-866
- Supplier No.:AM96SEP3100
Specifications
About this item
Silicone septa mat for capillary sequencing of 96-well PCR plates. Mat forms a tight seal on wells but is easily accessible through septa in each well. Ideal for work on ABI multi-capillary sequencing instruments.
Frequently Bought Together