ContactUsLinkComponent
Axymat* Sealing Mat
Axygen® Sequencing Septa Mats, Corning
Catalog # 47749-866
Supplier:  Corning
Axygen® Sequencing Septa Mats, Corning
Catalog # 47749-866
Supplier:  Corning
Specifications
  • Description:
    Sequencing Septa Mat
  • Material:
    Silicone
  • Cat. no.:
    47749-866
  • Supplier No.:
    AM96SEP3100
Specifications
About this item
Silicone septa mat for capillary sequencing of 96-well PCR plates. Mat forms a tight seal on wells but is easily accessible through septa in each well. Ideal for work on ABI multi-capillary sequencing instruments.
Frequently Bought Together