Spécifications
- Length:110 mm (41/5")
- Description:Tweezers Pattern 3C-SA
- Cat. no.:47743-168
- Tip Style:Straight, Ultra Fine
- Grade:3 Star
- Supplier No.:18056USA
Spécifications
A propos de cet article
Finely crafted tweezers
Tweezers 47743-180 and -182 feature flat edges with heavy-duty, strong blades tapering to regular strong points. Tweezers 47743-180 also has serrated tips and grips. Tweezers 47743-172 features angled fine tips that allow access into tight areas. Tweezers 47743-174 features curved super fine points for assembly work.
Tweezers 47743-178 features finger serrations and sturdy tips for bending and handling very fine wire and components. Tweezers 47743-168 features extra-long and narrow straight points for extended reach and access to confined areas. Tweezers 47743-170 is ideal for extra fine work, with long blades that break that sharply from body and taper to super-fine points.
Anti-glare finish to reduce eye strain. Hand finished points for accuracy. Recommended for micro electronics and precision assembly. Material: Stainless Steel, Anti-Magnetic, Anti-Acid.