ContactUsLinkComponent
Stainless Steel Precision Tweezers
Precision Stainless Steel Tweezers, Aven Tools
# de catalogue 47743-168
Fournisseur:  Aven Tools
Stainless Steel Precision Tweezers
Precision Stainless Steel Tweezers, Aven Tools
# de catalogue 47743-168
Fournisseur:  Aven Tools
Spécifications
  • Length:
    110 mm (41/5")
  • Description:
    Tweezers Pattern 3C-SA
  • Cat. no.:
    47743-168
  • Tip Style:
    Straight, Ultra Fine
  • Grade:
    3 Star
  • Supplier No.:
    18056USA
Spécifications
A propos de cet article
Finely crafted tweezers
Tweezers 47743-180 and -182 feature flat edges with heavy-duty, strong blades tapering to regular strong points. Tweezers 47743-180 also has serrated tips and grips. Tweezers 47743-172 features angled fine tips that allow access into tight areas. Tweezers 47743-174 features curved super fine points for assembly work.
Tweezers 47743-178 features finger serrations and sturdy tips for bending and handling very fine wire and components. Tweezers 47743-168 features extra-long and narrow straight points for extended reach and access to confined areas. Tweezers 47743-170 is ideal for extra fine work, with long blades that break that sharply from body and taper to super-fine points.
Anti-glare finish to reduce eye strain. Hand finished points for accuracy. Recommended for micro electronics and precision assembly. Material: Stainless Steel, Anti-Magnetic, Anti-Acid.
Customers Who Bought This Also Bought