KIMAX® Culture Tubes, Disposable, Soda-Lime Glass, Kimble Chase
# de catalogue 47735-728
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Capacity:6 ml
- Ø:12 mm
- Description:Plain Disposable Soda-Lime Glass Tubes
- Height:75 mm
- Supplier No.:60BM12
Spécifications
A propos de cet article
Plain disposable soda-lime glass tubes manufactured from 90 expansion soda-lime glass conforming to USP Type III requirements.
- All tubes have sturdy, uniform bottoms and consistent lengths
- No marking spot
Customers Who Bought This Also Bought