Axygen® Universal Bevelled Pipette Tips 200 µl, Corning
# de catalogue 47734-716
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Volume:200 µl
- Tip Color:Clear
- Sterile:+
- Supplier No.:T-200-C-STKS
Spécifications
A propos de cet article
These 200 µl tips are designed to fit a wide variety of single-channel and multi-channel pipettors.
Flexible walls and a series of internal sealing rings ensure a secure fit and less force required to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry. The bevelled tip reduces sample hang up and improves pipetting accuracy. Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures.
Packaging: 96 tips/rack, 5 racks/stack, 10 stacks/case.
Customers Who Bought This Also Bought