ContactUsLinkComponent
11mm Aluminum Seals
Seals for VWR® 11 mm Standard Opening Crimp-Top Vials, National Scientific
# de catalogue 46610-744
Fournisseur:  VWR International
11mm Aluminum Seals
Seals for VWR® 11 mm Standard Opening Crimp-Top Vials, National Scientific
# de catalogue 46610-744
Fournisseur:  VWR International
Spécifications
  • Description:
    Aluminum Seal with Clear PTFE/Natural Red Rubber Septa
  • Cat. no.:
    46610-744
  • Seal Size:
    11 mm
  • Cap Color:
    Silver